Product Details

SNP ID
rs147887185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55639174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGATAAAGTGTTTGCCCTTTTT[C/T]ATACCATCATTGCTCCCATGTTCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4P4 PubMed Links

Gene Details

Gene
OR4P4
Gene Name
olfactory receptor family 4 subfamily P member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004124.2 817 Missense Mutation CAT,TAT H273Y NP_001004124.1

View Full Product Details