Product Details

SNP ID
rs147949997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5389572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTCAGGCAATTGTTTCATTCT[C/G]ATCATTATTAAGACCAACCCTCGTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 196 Intron NP_001005567.2
Gene
OR51M1
Gene Name
olfactory receptor family 51 subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004756.2 196 Silent Mutation CTC,CTG L58L NP_001004756.2

View Full Product Details