Product Details

SNP ID
rs148012694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:110103089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTTTATTACAGCAGTCTGGAAC[C/T]AAAACCACAATATTTCCAAGGTATG
Phenotype
MIM: 615001
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZC3H12C PubMed Links

Gene Details

Gene
ZC3H12C
Gene Name
zinc finger CCCH-type containing 12C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033390.1 Intron NP_203748.1
XM_005271715.3 Intron XP_005271772.1
XM_011543055.2 Intron XP_011541357.1
XM_017018477.1 Intron XP_016873966.1

View Full Product Details