Product Details

SNP ID
rs148163879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62765665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCTCCTCAGGATATTGTGATT[C/T]AGCAGGACGATGAGATCCGCTTAAA
Phenotype
MIM: 602013 MIM: 602946
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLR2G PubMed Links

Gene Details

Gene
POLR2G
Gene Name
RNA polymerase II subunit G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002696.2 522 Nonsense Mutation CAG,TAG Q138* NP_002687.1
Gene
TAF6L
Gene Name
TATA-box binding protein associated factor 6 like
There are no transcripts associated with this gene.

View Full Product Details