Product Details

SNP ID
rs148276108
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66276056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCTTGTCAGGAGTTTGCAGAC[A/T]CTCTGGGCATCCCCTTCTTGGAGAC
Phenotype
MIM: 611288 MIM: 611729 MIM: 612565 MIM: 611484
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
CNIH2 PubMed Links

Gene Details

Gene
CNIH2
Gene Name
cornichon family AMPA receptor auxiliary protein 2
There are no transcripts associated with this gene.

Gene
KLC2
Gene Name
kinesin light chain 2
There are no transcripts associated with this gene.

Gene
RAB1B
Gene Name
RAB1B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030981.2 Intron NP_112243.1
XM_017018378.1 Intron XP_016873867.1
Gene
YIF1A
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
There are no transcripts associated with this gene.

View Full Product Details