Product Details

SNP ID
rs148294193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66468243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGGTGAAGATGCGCAGCCAGGC[C/G]AGGAGCCCATCAAGTATGGTGAACT
Phenotype
MIM: 609827
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928069 PubMed Links

Gene Details

Gene
LOC101928069
Gene Name
uncharacterized LOC101928069
There are no transcripts associated with this gene.

Gene
PELI3
Gene Name
pellino E3 ubiquitin protein ligase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098510.1 235 Missense Mutation CAG,GAG Q39E NP_001091980.1
NM_001243135.1 235 Missense Mutation CAG,GAG Q39E NP_001230064.1
NM_001243136.1 235 Intron NP_001230065.1
NM_145065.2 235 Missense Mutation CAG,GAG Q39E NP_659502.2
XM_011544884.2 235 Missense Mutation CAG,GAG Q39E XP_011543186.1
XM_017017465.1 235 UTR 5 XP_016872954.1

View Full Product Details