Product Details
- SNP ID
-
rs148529934
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:101453040 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTTCTGCTAGGTCTTCTGTATTC[C/T]GAGATTTTTCTTCAAGGAGTTCATA
- Phenotype
-
MIM: 603652
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TRPC6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12805398] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRPC6
- Gene Name
- transient receptor potential cation channel subfamily C member 6
View Full Product Details