Product Details

SNP ID
rs148589018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64318292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGGTGGGGCCGGCGGTCAGTCTG[C/T]GGCAGCGGCAGCAAGACGGTACAGT
Phenotype
MIM: 601998 MIM: 606583
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESRRA PubMed Links

Gene Details

Gene
ESRRA
Gene Name
estrogen related receptor alpha
There are no transcripts associated with this gene.

Gene
PRDX5
Gene Name
peroxiredoxin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012094.4 205 Missense Mutation GCG,GTG A26V NP_036226.1
NM_181651.2 205 Missense Mutation GCG,GTG A26V NP_857634.1
NM_181652.2 205 Missense Mutation GCG,GTG A26V NP_857635.1
XM_005273882.4 205 Missense Mutation GCG,GTG A26V XP_005273939.2
Gene
TRMT112
Gene Name
tRNA methyltransferase 11-2 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286082.1 205 Intron NP_001273011.1
NM_001286084.1 205 Intron NP_001273013.1
NM_016404.2 205 Intron NP_057488.1

View Full Product Details