Product Details

SNP ID
rs148686086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112078231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTCTAGATTCAGGAGGAATCAG[A/G]TATGTGGAAAATAAGAGAACTGGAA
Phenotype
MIM: 602690 MIM: 606659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf57 PubMed Links

Gene Details

Gene
C11orf57
Gene Name
chromosome 11 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082969.1 250 Missense Mutation GAT,GGT D29G NP_001076438.1
NM_001082970.1 250 Missense Mutation GAT,GGT D29G NP_001076439.1
NM_001301017.1 250 Missense Mutation GAT,GGT D29G NP_001287946.1
NM_001301019.1 250 UTR 5 NP_001287948.1
NM_001301021.1 250 UTR 5 NP_001287950.1
NM_018195.3 250 Missense Mutation GAT,GGT D29G NP_060665.3
Gene
PIH1D2
Gene Name
PIH1 domain containing 2
There are no transcripts associated with this gene.

Gene
SDHD
Gene Name
succinate dehydrogenase complex subunit D
There are no transcripts associated with this gene.

Gene
TIMM8B
Gene Name
translocase of inner mitochondrial membrane 8 homolog B
There are no transcripts associated with this gene.

View Full Product Details