Product Details

SNP ID
rs148794922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5884667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGTCATGGCTTATGACCGCTT[C/T]GTTGCCATCTGCAACCCTCTCCAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52E4 PubMed Links

Gene Details

Gene
OR52E4
Gene Name
olfactory receptor family 52 subfamily E member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005165.1 375 Silent Mutation TTC,TTT F125F NP_001005165.1

View Full Product Details