Product Details

SNP ID
rs148896304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126256697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAAATCTGACCTAGAACATTTCA[C/T]GCTGGTTCGGAGAGGAGACGTAGAT
Phenotype
MIM: 616587 MIM: 182180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM118B PubMed Links

Gene Details

Gene
FAM118B
Gene Name
family with sequence similarity 118 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024556.3 941 Missense Mutation ACG,ATG T276M NP_078832.1
XM_005271665.3 941 Missense Mutation ACG,ATG T276M XP_005271722.1
XM_011542977.2 941 Missense Mutation ACG,ATG T276M XP_011541279.1
XM_011542978.2 941 Missense Mutation ACG,ATG T121M XP_011541280.1
XM_017018285.1 941 Missense Mutation ACG,ATG T276M XP_016873774.1
XM_017018286.1 941 Missense Mutation ACG,ATG T121M XP_016873775.1
Gene
SRPRA
Gene Name
SRP receptor alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177842.1 941 Intron NP_001171313.1
NM_003139.3 941 Intron NP_003130.2
XM_017018179.1 941 UTR 3 XP_016873668.1

View Full Product Details