Product Details

SNP ID
rs149006752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72003982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGAGAACGGCGGGTTCCACTGC[A/G]AGTGTTGGGGGAAGCCTTGGACAGG
Phenotype
MIM: 604113 MIM: 164009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL18BP PubMed Links

Gene Details

Gene
IL18BP
Gene Name
interleukin 18 binding protein
There are no transcripts associated with this gene.

Gene
NUMA1
Gene Name
nuclear mitotic apparatus protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286561.1 6446 Missense Mutation CGC,TGC R2067C NP_001273490.1
NM_006185.3 6446 Missense Mutation CGC,TGC R2081C NP_006176.2
XM_006718564.1 6446 Missense Mutation CGC,TGC R2081C XP_006718627.1
XM_011545054.1 6446 Missense Mutation CGC,TGC R2099C XP_011543356.1
XM_011545055.1 6446 Missense Mutation CGC,TGC R2099C XP_011543357.1
XM_011545056.2 6446 Missense Mutation CGC,TGC R2099C XP_011543358.1
XM_011545057.2 6446 Missense Mutation CGC,TGC R2099C XP_011543359.1
XM_011545058.1 6446 Missense Mutation CGC,TGC R2099C XP_011543360.1
XM_011545059.2 6446 Missense Mutation CGC,TGC R2099C XP_011543361.1
XM_011545060.2 6446 Missense Mutation CGC,TGC R2099C XP_011543362.1
XM_011545061.2 6446 Missense Mutation CGC,TGC R2099C XP_011543363.1
XM_011545062.2 6446 Missense Mutation CGC,TGC R2099C XP_011543364.1
XM_011545063.2 6446 Missense Mutation CGC,TGC R2099C XP_011543365.1
XM_011545064.1 6446 Missense Mutation CGC,TGC R2099C XP_011543366.1
XM_011545065.1 6446 Missense Mutation CGC,TGC R2085C XP_011543367.1
XM_011545066.1 6446 Missense Mutation CGC,TGC R2067C XP_011543368.1
XM_017017830.1 6446 Missense Mutation CGC,TGC R2067C XP_016873319.1
XM_017017831.1 6446 Missense Mutation CGC,TGC R2067C XP_016873320.1
Gene
RNF121
Gene Name
ring finger protein 121
There are no transcripts associated with this gene.

View Full Product Details