Product Details

SNP ID
rs149221427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:14510890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTACCTTTGTAGTCAGGTCCTG[C/T]TCTGCAGGAAGCGTCTCTCTTAAGG
Phenotype
MIM: 602854
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMA1 PubMed Links

Gene Details

Gene
PSMA1
Gene Name
proteasome subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143937.1 752 Intron NP_001137409.1
NM_002786.3 752 Silent Mutation GAA,GAG E202E NP_002777.1
NM_148976.2 752 Silent Mutation GAA,GAG E208E NP_683877.1

View Full Product Details