Product Details

SNP ID
rs149382662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7796533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAACTACTAAGAGTAGCTGGACA[C/G]TGACTTGTGTGGACATTTTGGTTGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR5P2 PubMed Links

Gene Details

Gene
OR5P2
Gene Name
olfactory receptor family 5 subfamily P member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153444.1 410 Missense Mutation ACT,AGT T137S NP_703145.1

View Full Product Details