Product Details

SNP ID
rs149454771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68760252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAAAGCCCCCTCCGCTGGACAC[A/G]TACTCTGGGTTATTCTCCAAGTCAA
Phenotype
MIM: 600528 MIM: 604374
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPT1A PubMed Links

Gene Details

Gene
CPT1A
Gene Name
carnitine palmitoyltransferase 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031847.2 2285 Silent Mutation TAC,TAT Y705Y NP_001027017.1
NM_001876.3 2285 Silent Mutation TAC,TAT Y705Y NP_001867.2
XM_005273762.2 2285 Silent Mutation TAC,TAT Y737Y XP_005273819.1
XM_005273763.1 2285 Silent Mutation TAC,TAT Y737Y XP_005273820.1
XM_017017220.1 2285 Silent Mutation TAC,TAT Y705Y XP_016872709.1
Gene
TESMIN
Gene Name
testis expressed metallothionein like protein
There are no transcripts associated with this gene.

View Full Product Details