Product Details

SNP ID
rs150121776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:788439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGCTGGCGGCTGCTGCTTCTC[A/G]GCCGGGGCCTCCTTCTTCGAGGGCT
Phenotype
MIM: 608213 MIM: 609302
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEND1 PubMed Links

Gene Details

Gene
CEND1
Gene Name
cell cycle exit and neuronal differentiation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016564.3 317 Silent Mutation GCA,GCG A46A NP_057648.2
Gene
LOC171391
Gene Name
uncharacterized LOC171391
There are no transcripts associated with this gene.

Gene
PANO1
Gene Name
proapoptotic nucleolar protein 1
There are no transcripts associated with this gene.

Gene
SLC25A22
Gene Name
solute carrier family 25 member 22
There are no transcripts associated with this gene.

View Full Product Details