Product Details

SNP ID
rs150343311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5778059 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGCACAAGATAGCTTTACTAC[A/C]GACATGTGGTCGCAGTACGTATGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR52N1 PubMed Links

Gene Details

Gene
OR52N1
Gene Name
olfactory receptor family 52 subfamily N member 1
There are no transcripts associated with this gene.

Gene
OR52N5
Gene Name
olfactory receptor family 52 subfamily N member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001922.2 609 Silent Mutation TCG,TCT S192S NP_001001922.2

View Full Product Details