Product Details

SNP ID
rs137874227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48905010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAATAAGATTAACACACAGTGGA[C/G]AACTGTCCTTCGGGAAGTCAAGACC
Phenotype
MIM: 611088
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC65 PubMed Links

Gene Details

Gene
CCDC65
Gene Name
coiled-coil domain containing 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286957.1 424 UTR 5 NP_001273886.1
NM_033124.4 424 Missense Mutation ACA,AGA T66R NP_149115.2

View Full Product Details