Product Details

SNP ID
rs137882014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:45173582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTATCTTCTGGAGAGTGGTAAACT[A/C]TTCTTTGTTGGTTATATACTTCTGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLEKHA8P1 PubMed Links

Gene Details

Gene
PLEKHA8P1
Gene Name
pleckstrin homology domain containing A8 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details