Product Details

SNP ID
rs137981237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112013589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACACCAAGGGCGCCCTTCCCCTG[G/T]ATACGGTCACTTTCTACAAGGTAAC
Phenotype
MIM: 602287
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ERP29 PubMed Links

Gene Details

Gene
ERP29
Gene Name
endoplasmic reticulum protein 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034025.1 242 Missense Mutation GAT,TAT D42Y NP_001029197.1
NM_006817.3 242 Missense Mutation GAT,TAT D42Y NP_006808.1
XM_017018720.1 242 Intron XP_016874209.1
Gene
TMEM116
Gene Name
transmembrane protein 116
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193453.1 242 Intron NP_001180382.1
NM_001193531.1 242 Intron NP_001180460.1
NM_001294314.1 242 Intron NP_001281243.1
NM_138341.2 242 Intron NP_612350.1
XM_006719678.3 242 Intron XP_006719741.1
XM_011538946.1 242 Intron XP_011537248.1
XM_011538947.1 242 Intron XP_011537249.1
XM_011538948.2 242 Intron XP_011537250.1
XM_011538949.2 242 Intron XP_011537251.1
XM_011538950.1 242 Intron XP_011537252.1
XM_011538951.2 242 Intron XP_011537253.1
XM_011538953.1 242 Intron XP_011537255.1
XM_011538955.1 242 Intron XP_011537257.1
XM_011538957.1 242 Intron XP_011537259.1
XM_011538958.1 242 Intron XP_011537260.1
XM_017020187.1 242 Intron XP_016875676.1
XM_017020188.1 242 Intron XP_016875677.1
XM_017020189.1 242 Intron XP_016875678.1
XM_017020190.1 242 Intron XP_016875679.1
XM_017020191.1 242 Intron XP_016875680.1
XM_017020192.1 242 Intron XP_016875681.1
XM_017020193.1 242 Intron XP_016875682.1
XM_017020194.1 242 Intron XP_016875683.1

View Full Product Details