Product Details

SNP ID
rs138156966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10407699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCGAAGCACAGGCCAGCAAACT[C/T]TTTCTTCCCAAGTTCTTCTTTCCTT
Phenotype
MIM: 602892 MIM: 602893
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLRC3 PubMed Links

Gene Details

Gene
KLRC3
Gene Name
killer cell lectin like receptor C3
There are no transcripts associated with this gene.

Gene
KLRC4
Gene Name
killer cell lectin like receptor C4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013431.2 613 Missense Mutation AAA,AGA K144R NP_038459.1
Gene
KLRC4-KLRK1
Gene Name
KLRC4-KLRK1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199805.1 613 UTR 5 NP_001186734.1
Gene
LOC101928100
Gene Name
uncharacterized LOC101928100
There are no transcripts associated with this gene.

View Full Product Details