Product Details
- SNP ID
-
rs138186337
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:57694764 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTTTCCCAGAGCCAATCTTCGGA[C/T]GTGGTGATTGTCTCCTCTAAGTACA
- Phenotype
-
MIM: 609677
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OS9
PubMed Links
Gene Details
- Gene
- OS9
- Gene Name
- OS9, endoplasmic reticulum lectin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001017956.2 |
316 |
Silent Mutation |
GAC,GAT |
D59D |
NP_001017956.1 |
NM_001017957.2 |
316 |
Silent Mutation |
GAC,GAT |
D59D |
NP_001017957.1 |
NM_001017958.2 |
316 |
Silent Mutation |
GAC,GAT |
D59D |
NP_001017958.1 |
NM_001261420.1 |
316 |
Silent Mutation |
GAC,GAT |
D59D |
NP_001248349.1 |
NM_001261421.1 |
316 |
Silent Mutation |
GAC,GAT |
D59D |
NP_001248350.1 |
NM_001261422.1 |
316 |
Silent Mutation |
GAC,GAT |
D59D |
NP_001248351.1 |
NM_001261423.1 |
316 |
Intron |
|
|
NP_001248352.1 |
NM_006812.3 |
316 |
Silent Mutation |
GAC,GAT |
D59D |
NP_006803.1 |
XM_005268581.1 |
316 |
Silent Mutation |
GAC,GAT |
D59D |
XP_005268638.1 |
XM_006719200.1 |
316 |
Silent Mutation |
GAC,GAT |
D59D |
XP_006719263.1 |
XM_006719201.1 |
316 |
Silent Mutation |
GAC,GAT |
D59D |
XP_006719264.1 |
View Full Product Details