Product Details

SNP ID
rs138562955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123672124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGACTGGAGCGTGACTGTGATC[C/T]CCGGTGCGGTAAGGCCAGAAGTAAA
Phenotype
MIM: 601750 MIM: 613846
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTF2H3 PubMed Links

Gene Details

Gene
GTF2H3
Gene Name
general transcription factor IIH subunit 3
There are no transcripts associated with this gene.

Gene
TCTN2
Gene Name
tectonic family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143850.2 387 Missense Mutation CCC,TCC P87S NP_001137322.1
NM_024809.4 387 Missense Mutation CCC,TCC P87S NP_079085.2
XM_005253623.2 387 Missense Mutation CCC,TCC P87S XP_005253680.1
XM_006719605.3 387 Missense Mutation CCC,TCC P87S XP_006719668.1
XM_017019974.1 387 Missense Mutation CCC,TCC P87S XP_016875463.1
XM_017019975.1 387 UTR 5 XP_016875464.1

View Full Product Details