Product Details
- SNP ID
-
rs138988810
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52448859 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCATACCTGCTTCTTGACGTGGT[C/T]GATCTCAGATCTCAGCCTCTGGATC
- Phenotype
-
MIM: 148042
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT6B
PubMed Links
Gene Details
- Gene
- KRT6B
- Gene Name
- keratin 6B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005555.3 |
1234 |
Missense Mutation |
AAC,GAC |
N396D |
NP_005546.2 |
View Full Product Details