Product Details
- SNP ID
-
rs139613925
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:10409491 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTGTTTGGTTCCTGAAATGGAGA[C/T]TTTATTGCCCTTAAGTTTCCTTTGC
- Phenotype
-
MIM: 602892
MIM: 602893
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KLRC3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1841958] are located under a probe and SNP(s) [rs1841957] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLRC3
- Gene Name
- killer cell lectin like receptor C3
There are no transcripts associated with this gene.
- Gene
- KLRC4
- Gene Name
- killer cell lectin like receptor C4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013431.2 |
267 |
Missense Mutation |
ATC,GTC |
I29V |
NP_038459.1 |
- Gene
- KLRC4-KLRK1
- Gene Name
- KLRC4-KLRK1 readthrough
View Full Product Details