Product Details

SNP ID
rs140037467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103587515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAGTAATTTTTTGTCTTGGATT[G/T]GTTGTACAAAATTTCTGCTCCCCAG
Phenotype
MIM: 608561
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
STAB2 PubMed Links

Gene Details

Gene
STAB2
Gene Name
stabilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017564.9 266 Silent Mutation TTG,TTT L13F NP_060034.9
XM_011538537.2 266 Silent Mutation TTG,TTT L13F XP_011536839.1
XM_011538538.2 266 Silent Mutation TTG,TTT L13F XP_011536840.1
XM_011538539.2 266 Intron XP_011536841.1
XM_011538541.2 266 Intron XP_011536843.1
XM_011538542.2 266 Intron XP_011536844.1
XM_017019585.1 266 Silent Mutation TTG,TTT L13F XP_016875074.1

View Full Product Details