Product Details

SNP ID
rs140348502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103951745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAACAAAAAGTTCCCTTGCTTGC[C/T]ACTCAATGATAACTCTTTATTATGA
Phenotype
MIM: 191175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf73 PubMed Links

Gene Details

Gene
C12orf73
Gene Name
chromosome 12 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135570.1 Intron NP_001129042.1
XM_011538718.2 Intron XP_011537020.1
XM_017019916.1 Intron XP_016875405.1
XM_017019917.1 Intron XP_016875406.1
Gene
HSP90B1
Gene Name
heat shock protein 90 beta family member 1
There are no transcripts associated with this gene.

View Full Product Details