Product Details

SNP ID
rs140431274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56131514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCGGGTGATACTGGAGCCACTCA[C/T]TGGGGACCTTCCCTTCGTGGGGGCT
Phenotype
MIM: 616670
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESYT1 PubMed Links

Gene Details

Gene
ESYT1
Gene Name
extended synaptotagmin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184796.1 870 Missense Mutation ACT,ATT T251I NP_001171725.1
NM_015292.2 870 Missense Mutation ACT,ATT T251I NP_056107.1
Gene
ZC3H10
Gene Name
zinc finger CCCH-type containing 10
There are no transcripts associated with this gene.

View Full Product Details