Product Details

SNP ID
rs140534652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53051883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAAGCTGAGCCTCATAGCTTCC[A/G]GGAGAAGGTTTTCCGGAAGAAACCT
Phenotype
MIM: 603928 MIM: 607717
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4B PubMed Links

Gene Details

Gene
EIF4B
Gene Name
eukaryotic translation initiation factor 4B
There are no transcripts associated with this gene.

Gene
LOC283335
Gene Name
uncharacterized LOC283335
There are no transcripts associated with this gene.

Gene
MIR6757
Gene Name
microRNA 6757
There are no transcripts associated with this gene.

Gene
TNS2
Gene Name
tensin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015319.2 130 Missense Mutation CAG,CGG Q45R NP_056134.2
NM_170754.2 130 Missense Mutation CAG,CGG Q35R NP_736610.2
NM_198316.1 130 UTR 5 NP_938072.1
XM_006719302.3 130 Missense Mutation CAG,CGG Q45R XP_006719365.1
XM_006719303.1 130 Missense Mutation CAG,CGG Q35R XP_006719366.1
XM_011538079.1 130 Missense Mutation CAG,CGG Q29R XP_011536381.1
XM_017019088.1 130 Missense Mutation CAG,CGG Q35R XP_016874577.1
XM_017019089.1 130 Missense Mutation CAG,CGG Q29R XP_016874578.1

View Full Product Details