Product Details

SNP ID
rs141592108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:15624257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAATCCCTTTGACTGTAACCAC[A/G]TCTTCACATCCTCTGGTGTGGAGTC
Phenotype
MIM: 600206
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPS8 PubMed Links

Gene Details

Gene
EPS8
Gene Name
epidermal growth factor receptor pathway substrate 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004447.5 2692 Missense Mutation ACG,ATG T732M NP_004438.3
XM_011520605.2 2692 Missense Mutation ACG,ATG T752M XP_011518907.1

View Full Product Details