Product Details

SNP ID
rs141632719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56010982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTATTAGGGACTTTTGGGCACAA[C/T]TGCAATCAAAGGCAATTGTTCCTGT
Phenotype
MIM: 606239 MIM: 606887
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IKZF4 PubMed Links

Gene Details

Gene
IKZF4
Gene Name
IKAROS family zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022465.3 Intron NP_071910.3
XM_005269086.3 Intron XP_005269143.1
XM_005269089.2 Intron XP_005269146.1
XM_005269090.3 Intron XP_005269147.1
XM_005269093.2 Intron XP_005269150.1
XM_011538664.1 Intron XP_011536966.1
XM_011538666.2 Intron XP_011536968.1
XM_011538669.2 Intron XP_011536971.1
XM_017019805.1 Intron XP_016875294.1
XM_017019806.1 Intron XP_016875295.1
XM_017019807.1 Intron XP_016875296.1
XM_017019808.1 Intron XP_016875297.1
XM_017019809.1 Intron XP_016875298.1
XM_017019810.1 Intron XP_016875299.1
XM_017019811.1 Intron XP_016875300.1
XM_017019812.1 Intron XP_016875301.1
XM_017019813.1 Intron XP_016875302.1
XM_017019814.1 Intron XP_016875303.1
XM_017019815.1 Intron XP_016875304.1
XM_017019816.1 Intron XP_016875305.1
Gene
LOC105369781
Gene Name
uncharacterized LOC105369781
There are no transcripts associated with this gene.

Gene
SUOX
Gene Name
sulfite oxidase
There are no transcripts associated with this gene.

View Full Product Details