Product Details

SNP ID
rs141755106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7479959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAAATTTAGACACAGAAATTAG[C/T]TCAGCAGCACTGAAATCAGCTGACT
Phenotype
MIM: 605545
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD163 PubMed Links

Gene Details

Gene
CD163
Gene Name
CD163 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004244.5 3403 Silent Mutation GAA,GAG E1127E NP_004235.4
NM_203416.3 3403 Intron NP_981961.2
XM_005253528.4 3403 Intron XP_005253585.1
XM_005253529.4 3403 Silent Mutation GAA,GAG E1095E XP_005253586.1
XM_017020231.1 3403 Intron XP_016875720.1

View Full Product Details