Product Details

SNP ID
rs141771500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101698121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGTGCTCATCTCCTACTGTCCCA[C/T]GGCCACCGAAGAGGTAGGGCTGGCC
Phenotype
MIM: 616747 MIM: 160794
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHPT1 PubMed Links

Gene Details

Gene
CHPT1
Gene Name
choline phosphotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020244.2 483 Missense Mutation ACG,ATG T87M NP_064629.2
XM_011538574.1 483 Missense Mutation ACG,ATG T87M XP_011536876.1
XM_011538575.1 483 Intron XP_011536877.1
Gene
MYBPC1
Gene Name
myosin binding protein C, slow type
There are no transcripts associated with this gene.

View Full Product Details