Product Details
- SNP ID
-
rs142318174
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:120978923 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGACAAGGGGGAGTCCTGCGGCG[C/G]CGGTCGAGGGGAGCTGGCTGAGCTG
- Phenotype
-
MIM: 142410
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HNF1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1169288] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HNF1A
- Gene Name
- HNF1 homeobox A
- Gene
- HNF1A-AS1
- Gene Name
- HNF1A antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details