Product Details

SNP ID
rs142483430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122172678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGATGTTGTTTGGCGGCTGGG[C/T]GTCAGGGCTGAGACACGGCAGCGTG
Phenotype
MIM: 609509
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL31 PubMed Links

Gene Details

Gene
IL31
Gene Name
interleukin 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014336.1 256 Missense Mutation ACC,GCC T77A NP_001014358.1
Gene
LRRC43
Gene Name
leucine rich repeat containing 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098519.1 256 Intron NP_001091989.1
NM_152759.4 256 Intron NP_689972.3
XM_017019124.1 256 Intron XP_016874613.1
XM_017019125.1 256 Intron XP_016874614.1

View Full Product Details