Product Details

SNP ID
rs142523671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55130022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCTCGGCTGAACACACCAATGT[A/G]TTTTTTCCTAGGCAATCTCTCCTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR9K2 PubMed Links

Gene Details

Gene
OR9K2
Gene Name
olfactory receptor family 9 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005243.1 254 Missense Mutation TAT,TGT Y85C NP_001005243.1

View Full Product Details