Product Details

SNP ID
rs142606266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:61708782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATTTTACATTTTTAGTTGCTCTT[A/C]CCAGGAAAGTGTCTGAGTAGTGTTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM19A2 PubMed Links

Gene Details

Gene
FAM19A2
Gene Name
family with sequence similarity 19 member A2, C-C motif chemokine like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178539.4 3602 UTR 3 NP_848634.1
XM_006719370.2 3602 UTR 3 XP_006719433.1
XM_011538272.1 3602 UTR 3 XP_011536574.1
XM_011538274.1 3602 UTR 3 XP_011536576.1

View Full Product Details