Product Details

SNP ID
rs142930741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55721481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGCCAAGTGGGTGGAGATGCA[C/T]GTTAAGGAAGCAGGTAAGTATGGTA
Phenotype
MIM: 601444 MIM: 155740 MIM: 600536 MIM: 601617
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BLOC1S1 PubMed Links

Gene Details

Gene
BLOC1S1
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 1
There are no transcripts associated with this gene.

Gene
BLOC1S1-RDH5
Gene Name
BLOC1S1-RDH5 readthrough
There are no transcripts associated with this gene.

Gene
CD63
Gene Name
CD63 molecule
There are no transcripts associated with this gene.

Gene
ITGA7
Gene Name
integrin subunit alpha 7
There are no transcripts associated with this gene.

Gene
RDH5
Gene Name
retinol dehydrogenase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199771.1 480 Silent Mutation CAC,CAT H99H NP_001186700.1
NM_002905.3 480 Silent Mutation CAC,CAT H99H NP_002896.2

View Full Product Details