Product Details

SNP ID
rs142985348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:65281400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTATGTAACTCTTCAGAGCCA[C/T]CTACTACATGTGGTTCAAACTTCTG
Phenotype
MIM: 613719
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSRB3 PubMed Links

Gene Details

Gene
MSRB3
Gene Name
methionine sulfoxide reductase B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031679.2 Intron NP_001026849.1
NM_001193460.1 Intron NP_001180389.1
NM_001193461.1 Intron NP_001180390.1
NM_198080.3 Intron NP_932346.1

View Full Product Details