Product Details

SNP ID
rs143015363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56995307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATCCACAACTGGACCGAGCTGC[C/T]TGACCTCTTCAACCACACTTTGTCT
Phenotype
MIM: 605307
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR182 PubMed Links

Gene Details

Gene
GPR182
Gene Name
G protein-coupled receptor 182
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007264.3 868 Missense Mutation CCT,CTT P33L NP_009195.1
XM_011537824.2 868 Missense Mutation CCT,CTT P33L XP_011536126.1
XM_011537825.2 868 Missense Mutation CCT,CTT P33L XP_011536127.1
XM_011537827.2 868 Missense Mutation CCT,CTT P33L XP_011536129.1
Gene
HBCBP
Gene Name
HBcAg-binding protein
There are no transcripts associated with this gene.

Gene
ZBTB39
Gene Name
zinc finger and BTB domain containing 39
There are no transcripts associated with this gene.

View Full Product Details