Product Details

SNP ID
rs143213723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55221173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTTCTTATTGTCACAGTTACCA[A/G]TGTGGATCTCGCACTTCAAACACCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10A7 PubMed Links

Gene Details

Gene
OR10A7
Gene Name
olfactory receptor family 10 subfamily A member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005280.1 149 Missense Mutation AAT,AGT N50S NP_001005280.1

View Full Product Details