Product Details
- SNP ID
-
rs143644838
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:110013231 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCCCTTGAGGGAGTTCCTGAGCT[A/G]CTCCAAAAAATTCTCGAGGTATCCA
- Phenotype
-
MIM: 615123
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD13A
PubMed Links
Gene Details
- Gene
- ANKRD13A
- Gene Name
- ankyrin repeat domain 13A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033121.1 |
595 |
Silent Mutation |
CTA,CTG |
L112L |
NP_149112.1 |
XM_005253980.2 |
595 |
Silent Mutation |
CTA,CTG |
L112L |
XP_005254037.1 |
XM_005253981.3 |
595 |
Silent Mutation |
CTA,CTG |
L112L |
XP_005254038.1 |
XM_005253982.3 |
595 |
Silent Mutation |
CTA,CTG |
L112L |
XP_005254039.1 |
XM_005253984.1 |
595 |
Silent Mutation |
CTA,CTG |
L24L |
XP_005254041.1 |
XM_011538937.1 |
595 |
Silent Mutation |
CTA,CTG |
L112L |
XP_011537239.1 |
XM_011538938.2 |
595 |
Silent Mutation |
CTA,CTG |
L112L |
XP_011537240.1 |
XM_017020157.1 |
595 |
Silent Mutation |
CTA,CTG |
L112L |
XP_016875646.1 |
XM_017020158.1 |
595 |
Silent Mutation |
CTA,CTG |
L112L |
XP_016875647.1 |
XM_017020159.1 |
595 |
Silent Mutation |
CTA,CTG |
L24L |
XP_016875648.1 |
XM_017020160.1 |
595 |
Silent Mutation |
CTA,CTG |
L24L |
XP_016875649.1 |
XM_017020161.1 |
595 |
Silent Mutation |
CTA,CTG |
L24L |
XP_016875650.1 |
XM_017020162.1 |
595 |
Silent Mutation |
CTA,CTG |
L112L |
XP_016875651.1 |
XM_017020163.1 |
595 |
UTR 5 |
|
|
XP_016875652.1 |
View Full Product Details