Product Details

SNP ID
rs144147963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:16550514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTATTCTAAAAATGGCACTTTTCA[C/T]GTTGGTATAAAATGAAAGTGCTTTA
Phenotype
MIM: 180386
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMO3 PubMed Links

Gene Details

Gene
LMO3
Gene Name
LIM domain only 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001395.2 1500 UTR 3 NP_001001395.1
NM_001243609.1 1500 UTR 3 NP_001230538.1
NM_001243610.1 1500 UTR 3 NP_001230539.1
NM_001243611.1 1500 UTR 3 NP_001230540.1
NM_001243612.1 1500 UTR 3 NP_001230541.1
NM_001243613.1 1500 UTR 3 NP_001230542.1
NM_018640.4 1500 UTR 3 NP_061110.2
XM_006719110.3 1500 UTR 3 XP_006719173.1
XM_006719111.2 1500 UTR 3 XP_006719174.1
XM_011520762.1 1500 UTR 3 XP_011519064.1
XM_011520763.1 1500 UTR 3 XP_011519065.1
XM_017019643.1 1500 UTR 3 XP_016875132.1

View Full Product Details