Product Details

SNP ID
rs144443479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57434972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTGGCGGTCAGTGTCCAGCAT[A/G]TGGGGGGCCCACCTTGGAACTGGAG
Phenotype
MIM: 601233
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INHBC PubMed Links

Gene Details

Gene
INHBC
Gene Name
inhibin beta C subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005538.3 288 Missense Mutation TAT,TGT Y29C NP_005529.1
Gene
R3HDM2
Gene Name
R3H domain containing 2
There are no transcripts associated with this gene.

View Full Product Details