Product Details

SNP ID
rs144754856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:94149136 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCATCGCGGCGAGCCAGGAGGA[C/T]GGCGTGTTTGTGGCGAGCGGCAGCT
Phenotype
MIM: 604259
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLXNC1 PubMed Links

Gene Details

Gene
PLXNC1
Gene Name
plexin C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005761.2 441 Silent Mutation GAC,GAT D55D NP_005752.1
XM_006719186.3 441 Silent Mutation GAC,GAT D55D XP_006719249.1
XM_011537730.2 441 Silent Mutation GAC,GAT D55D XP_011536032.1
XM_011537731.2 441 Silent Mutation GAC,GAT D55D XP_011536033.1
XM_017018671.1 441 Silent Mutation GAC,GAT D55D XP_016874160.1
XM_017018672.1 441 Silent Mutation GAC,GAT D55D XP_016874161.1

View Full Product Details