Product Details
- SNP ID
-
rs145186457
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:105174376 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCCATCATCGTCATCTGGTTGAT[C/G]GTTTAACAGTACATATTTTCCGTCA
- Phenotype
-
MIM: 606231
MIM: 615748
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
APPL2
PubMed Links
Gene Details
- Gene
- APPL2
- Gene Name
- adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001251904.1 |
3266 |
Missense Mutation |
CAT,GAT |
H651D |
NP_001238833.1 |
NM_001251905.1 |
3266 |
Missense Mutation |
CAT,GAT |
H602D |
NP_001238834.1 |
NM_018171.3 |
3266 |
Missense Mutation |
CAT,GAT |
H645D |
NP_060641.2 |
XM_006719472.1 |
3266 |
Missense Mutation |
CAT,GAT |
H604D |
XP_006719535.1 |
XM_011538530.2 |
3266 |
Missense Mutation |
CAT,GAT |
H638D |
XP_011536832.1 |
XM_011538531.2 |
3266 |
Missense Mutation |
CAT,GAT |
H608D |
XP_011536833.1 |
XM_011538532.2 |
3266 |
Missense Mutation |
CAT,GAT |
H608D |
XP_011536834.1 |
XM_017019551.1 |
3266 |
Missense Mutation |
CAT,GAT |
H632D |
XP_016875040.1 |
XM_017019552.1 |
3266 |
Missense Mutation |
CAT,GAT |
H602D |
XP_016875041.1 |
XM_017019553.1 |
3266 |
Missense Mutation |
CAT,GAT |
H602D |
XP_016875042.1 |
XM_017019554.1 |
3266 |
Missense Mutation |
CAT,GAT |
H598D |
XP_016875043.1 |
- Gene
- KIAA1033
- Gene Name
- KIAA1033
There are no transcripts associated with this gene.
View Full Product Details