Product Details

SNP ID
rs145629853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:63844846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCCCGAGCCGATTCAAGAAGGA[C/G]AAAGAGATCATAGCCGAGTATGAAA
Phenotype
MIM: 606523
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SRGAP1 PubMed Links

Gene Details

Gene
SRGAP1
Gene Name
SLIT-ROBO Rho GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020762.2 40 Missense Mutation GAC,GAG D10E NP_065813.1
XM_005269042.3 40 Missense Mutation GAC,GAG D10E XP_005269099.1
XM_011538580.1 40 UTR 5 XP_011536882.1
XM_011538581.2 40 Intron XP_011536883.1

View Full Product Details