Product Details

SNP ID
rs146045476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54396363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGTGCCATATGGGAGGGAGCGT[C/T]TGAAGAATCCAAGCTGATAAAGGGG
Phenotype
MIM: 135620 MIM: 609124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGA5 PubMed Links

Gene Details

Gene
ITGA5
Gene Name
integrin subunit alpha 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002205.4 3148 Missense Mutation AAA,AGA K1027R NP_002196.4
Gene
LOC102724050
Gene Name
uncharacterized LOC102724050
There are no transcripts associated with this gene.

Gene
ZNF385A
Gene Name
zinc finger protein 385A
There are no transcripts associated with this gene.

View Full Product Details