Product Details

SNP ID
rs146077019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21471630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAGATCTCTGCAGTACTCTGTTA[C/T]GTTCTTTCTTTCAAATGCTGTAATA
Phenotype
MIM: 600537
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PYROXD1 PubMed Links

Gene Details

Gene
PYROXD1
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024854.3 1884 Intron NP_079130.2
XM_006719153.3 1884 Intron XP_006719216.1
XM_017019976.1 1884 Intron XP_016875465.1
Gene
RECQL
Gene Name
RecQ like helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002907.3 1884 Missense Mutation ATA,GTA I489V NP_002898.2
NM_032941.2 1884 Missense Mutation ATA,GTA I489V NP_116559.1
XM_005253461.3 1884 Missense Mutation ATA,GTA I489V XP_005253518.1
XM_005253462.4 1884 Missense Mutation ATA,GTA I489V XP_005253519.1
XM_005253463.3 1884 Missense Mutation ATA,GTA I489V XP_005253520.1
XM_005253464.3 1884 Missense Mutation ATA,GTA I489V XP_005253521.1

View Full Product Details