Product Details

SNP ID
rs146297198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:25108768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTGTGCTCTGTCACCACTAAAC[A/G]TTAAAAGGGCCCAATTAGGATTCTC
Phenotype
MIM: 616906 MIM: 602003
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASC1 PubMed Links

Gene Details

Gene
CASC1
Gene Name
cancer susceptibility candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082972.2 2112 Intron NP_001076441.1
NM_001082973.2 2112 Missense Mutation NP_001076442.1
NM_001204101.2 2112 Missense Mutation NP_001191030.1
NM_001204102.2 2112 Missense Mutation NP_001191031.1
NM_001319977.1 2112 Intron NP_001306906.1
NM_001319978.1 2112 Missense Mutation NP_001306907.1
NM_018272.4 2112 Missense Mutation NP_060742.3
XM_005253406.1 2112 Missense Mutation XP_005253463.1
XM_005253408.1 2112 Missense Mutation XP_005253465.1
XM_005253409.1 2112 Missense Mutation XP_005253466.1
XM_006719101.1 2112 Missense Mutation XP_006719164.1
XM_011520723.1 2112 Missense Mutation XP_011519025.1
XM_011520726.1 2112 Intron XP_011519028.1
XM_011520727.1 2112 Intron XP_011519029.1
XM_017019560.1 2112 Missense Mutation XP_016875049.1
XM_017019561.1 2112 Missense Mutation XP_016875050.1
XM_017019562.1 2112 Intron XP_016875051.1
XM_017019563.1 2112 Missense Mutation XP_016875052.1
XM_017019564.1 2112 Missense Mutation XP_016875053.1
XM_017019565.1 2112 Intron XP_016875054.1
XM_017019566.1 2112 Missense Mutation XP_016875055.1
XM_017019567.1 2112 Missense Mutation XP_016875056.1
XM_017019568.1 2112 Intron XP_016875057.1
Gene
LRMP
Gene Name
lymphoid restricted membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204126.1 2112 Intron NP_001191055.1
NM_001204127.1 2112 Intron NP_001191056.1
NM_006152.3 2112 Intron NP_006143.2
XM_005253370.3 2112 Intron XP_005253427.1
XM_005253372.2 2112 Intron XP_005253429.1
XM_005253374.3 2112 Intron XP_005253431.1
XM_006719076.2 2112 Intron XP_006719139.1
XM_011520667.1 2112 Intron XP_011518969.1
XM_011520668.2 2112 Intron XP_011518970.1
XM_011520669.2 2112 Intron XP_011518971.1
XM_011520670.1 2112 Intron XP_011518972.1
XM_017019300.1 2112 Intron XP_016874789.1
XM_017019301.1 2112 Intron XP_016874790.1
XM_017019302.1 2112 Intron XP_016874791.1

View Full Product Details